Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1402046

Sigma-Aldrich

Anti-PTPLB antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

3-Hydroxyacyl-CoA dehydratase 2, HACD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PTPLB(201562)

General description

Protein tyrosine phosphatase-like member b gene (PTPLB) is an integral protein of endoplasmic reticulum, with four membrane-spanning alpha helices. PTPLB is also known as 3-hydroxyacyl-CoA dehydratase 2 (HACD2).

Immunogen

PTPLB (NP_940684.1, 1 a.a. ~ 254 a.a) full-length human protein.

Sequence
MAAVAATAAAKGNGGGGGRAGAGDASGTRKKKGPGPLATAYLVIYNVVMTAGWLVIAVGLVRAYLAKGSYHSLYYSIEKPLKFFQTGALLEILHCAIGIVPSSVVLTSFQVMSRVFLIWAVTHSVKEVQSEDSVLLFVIAWTITEIIRYSFYTFSLLNHLPYLIKWARYTLFIVLYPMGVSGELLTIYAALPFVRQAGLYSISLPNKYNFSFDYYAFLILIMISYIPIFPQLYFHMIHQRRKILSHTEEHKKFE

Biochem/physiol Actions

Protein tyrosine phosphatase-like member b gene (PTPLB) catalyses the third step in the elongation of fatty acid synthesis. It catalyses the dehydration of the 3-hydroxyacyl-CoA.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The role of fatty acid elongation in epidermal structure and function
Uchida and Yoshikazu.
Dermato-endocrinology, 3(2) (2011)
The 3-hydroxyacyl-CoA dehydratases HACD1 and HACD2 exhibit functional redundancy and are active in a wide range of fatty acid elongation pathways
Sawai, et al.
The Journal of Biological Chemistry (2017)
The yeast split-ubiquitin membrane protein two-hybrid screen identifies BAP31 as a regulator of the turnover of endoplasmic reticulum-associated protein tyrosine phosphatase-like B
Wang, et al.
Molecular and Cellular Biology, 24(7) (2004)
Mika Ikeda et al.
FEBS letters, 582(16), 2435-2440 (2008-06-17)
Very long-chain fatty acids are produced through a four-step cycle. However, the 3-hydroxyacyl-CoA dehydratase catalyzing the third step in mammals has remained unidentified. Mammals have four candidates, HACD1-4, based on sequence similarities to the recently identified yeast Phs1, although HACD3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service