Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1401830

Sigma-Aldrich

Anti-SNIP1 antibody produced in rabbit

whole antiserum, buffered aqueous solution

Synonym(s):

FLJ12553, RP3-423B22.3, dJ423B22.2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNIP1(79753)

Related Categories

Immunogen

SNIP1 (NP_078976.2, 1 a.a. ~ 396 a.a) full-length human protein.

Sequence
MKAVKSERERGSRRRHRDGDVVLPAGVVVKQERLSPEVAPPAHRRPDHSGGSPSPPTSEPARSGHRGNRARGVSRSPPKKKNKASGRRSKSPRSKRNRSPHHSTVKVKQEREDHPRRGREDRQHREPSEQEHRRARNSDRDRHRGHSHQRRTSNERPGSGQGQGRDRDTQNLQAQEEEREFYNARRREHRQRNDVGGGGSESQELVPRPGGNNKEKEVPAKEKPSFELSGALLEDTNTFRGVVIKYSEPPEARIPKKRWRLYPFKNDEVLPVMYIHRQSAYLLGRHRRIADIPIDHPSCSKQHAVFQYRLVEYTRADGTVGRRVKPYIIDLGSGNGTFLNNKRIEPQRYYELKEKDVLKFGFSSREYVLLHESSDTSEIDRKDDEDEEEEEEVSDS

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service