Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401769

Sigma-Aldrich

Anti-CFC1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRYPTIC, FLJ77897, HTX2, MGC133213

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CFC1(55997)

General description

This gene encodes a member of the epidermal growth factor (EGF)- Cripto, Frl-1, and Cryptic (CFC) family. EGF-CFC family member proteins share a variant EGF-like motif, a conserved cysteine-rich domain, and a C-terminal hydrophobic region. These proteins play key roles in intercellular signaling pathways during vertebrate embryogenesis. Mutations in this gene can cause autosomal visceral heterotaxy. This protein is involved in left-right asymmetric morphogenesis during organ development. (provided by RefSeq)

Immunogen

CFC1 (NP_115934.1, 1 a.a. ~ 223 a.a) full-length human protein.

Sequence
MTWRHHVRLLFTVSLALQIINLGNSYQREKHNGGREEVTKVATQKHRQSPLNWTSSHFGEVTGSAEGWGPEEPLPYSRAFGEGASARPRCCRNGGTCVLGSFCVCPAHFTGRYCEHDQRRSECGALEHGAWTLRACHLCRCIFGALHCLPLQTPDRCDPKDFLASHAHGPSAGGAPSLLLLLPCALLHRLLRPDAPAHPRSLVPSVLQRERRPCGRPGLGHRL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service