Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401706

Sigma-Aldrich

Monoclonal Anti-ASB2 antibody produced in mouse

clone 1E5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

ASB-2, MGC40044

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ASB2(51676)

Related Categories

General description

The protein encoded by this gene is a member of the ankyrin repeat and SOCS box-containing (ASB) family of proteins. They contain ankyrin repeat sequence and SOCS box domain. The SOCS box serves to couple suppressor of cytokine signalling (SOCS) proteins and their binding partners with the elongin B and C complex, possibly targeting them for degradation. This gene is induced by all-trans retinoic acid. In myeloid leukemia cells, the expression of this encoded protein has been shown to induce growth inhibition and chromatin condensation. Multiple alternatively spliced transcript variants have been described for this gene but their full length sequences are not known. (provided by RefSeq)

Immunogen

ASB2 (AAH32354, 1 a.a. ~ 587 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MTRFSYAEYFSLFHSCSAPSRSTAPPESSPARAPMGSFQGVMQKYSSSLFKTSQLAPADPLIKAIKDGDEEALKTMIKEGKNLAEPNKEGWLPLHEAAYYGQVGCLKVLQRAYPGTIDQRTLQEETAVYLATCRGHLDCLLSLLQAGAEPDISNKSRETPLYKACERKNAEAVKILVQHNADTNHRCNRGWTALHESVSRNDLEVMQILVSGGAKVESKNAYGITPLFVAAQSGQLEALRFLAKYGADINTQASDNASALYEACKNEHEEVVEFLLSQGADANKTNKDGLLPLHIASKKGNYRIVQMLLPVTSRTRIRRSGVSPLHLAAERNHDEVLEALLSARFDVNTPLAPERARLYEDRRSSALYFAVVNNNVYATELLLQHGADPNRDVISPLLVAIRHGCLRTMQLLLDHGANIDAYIATHPTAFPATIMFAMKCLSLLKFLMDLGCDGEPCFSCLYGNGPHPPAPQPSSRFNDAPAADKEPSVVQFCEFVSAPEVSRWAGPIIDVLLDYVGNVQLCSRLKEHIDSFEDWAVIKEKAEPPRPLAHLCRLRVRKAIGKYRIKLLDTLPLPGRLIRYLKYENTQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service