Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401703

Sigma-Aldrich

Anti-TAF9B antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

DN-7, DN7, TAF9L, TAFII31L, TFIID-31

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAF9B(51616)

Related Categories

General description

Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals. TFIID is composed of the TATA-binding protein (TBP) and a group of evolutionarily conserved proteins known as TBP-associated factors or TAFs. TAFs may participate in basal transcription, serve as coactivators, function in promoter recognition or modify general transcription factors (GTFs) to facilitate complex assembly and transcription initiation. This gene encodes a protein that is similar to one of the small subunits of TFIID, TBP-associated factor 9, and is also a subunit of TFIID. TAF9 and TAF9b share some functions but also have distinct roles in the transcriptional regulatory process. (provided by RefSeq)

Immunogen

TAF9B (NP_057059.2, 1 a.a. ~ 251 a.a) full-length human protein.

Sequence
MESGKMAPPKNAPRDALVMAQILKDMGITEYEPRVINQMLEFAFRYVTTILDDAKIYSSHAKKPNVDADDVRLAIQCRADQSFTSPPPRDFLLDIARQKNQTPLPLIKPYAGPRLPPDRYCLTAPNYRLKSLIKKGPNQGRLVPRLSVGAVSSKPTTPTIATPQTVSVPNKVATPMSVTSQRFTVQIPPSQSTPVKPVPATTAVQNVLINPSMIGPKNILITTNMVSSQNTANEANPLKRKHEDDDDNDIM

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service