Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401674

Sigma-Aldrich

Monoclonal Anti-MRPL4 antibody produced in mouse

clone 4B1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CGI-28, L4mt, MGC16367, MGC2681

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4B1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

capture ELISA: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPL4(51073)

General description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. Sequence analysis identified alternatively spliced variants that encode different protein isoforms. (provided by RefSeq)

Immunogen

MRPL4 (AAH00756, 1 a.a. ~ 263 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MLQFVRAGARAWLRPTGSQGLSSLAEEAARATENPEQVASEGLPEPVLRKVELPVPTHRRPVQAWVESLRGFEQERVGLADLHPDVFATAPRLDILHQVAMWQKNFKRISYAKTKTRAEVRGGGRKPWPQKGTGRARHGSIRSPLWRGGGVAHGPRGPTSYYYMLPMKVRALGLKVALTVKLAQDDLHIMDSLELPTGDPQYLTELAHYRRWGDSVLLVDLTHEEMPQSIVEATSRLKTFNLIPAVGEQRAQAPRVRMCRLRC

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service