Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401460

Sigma-Aldrich

Anti-GPR50 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

H9, MGC125342, Mel1c

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... GPR50(9248)

Immunogen

GPR50 (AAI03697.1, 1 a.a. ~ 617 a.a) full-length human protein.

Sequence
MGPTLAVPTPYGCIGCKLPQPEYPPALIIFMFCAMVITIVVDLIGNSMVILAVTKNKKLRNSGNIFVVSLSVADMLVAIYPYPLMLHAMSIGGWDLSQLQCQMVGFITGLSVVGSIFNIVAIAINRYCYICHSLQYERIFSVRNTCIYLVITWIMTVLAVLPNMYIGTIEYDPRTYTCIFNYLNNPVFTVTIVCIHFVLPLLIVGFCYVRIWTKVLAARDPAGQNPDNQLAEVRNFLTMFVIFLLFAVCWCPINVLTVLVAVSPKEMAGKIPNWLYLAAYFIAYFNSCLNAVIYGLLNENFRREYWTIFHAMRHPIIFFSGLISDIREMQEARTLARARAHARDQAREQDRAHACPAVEETPMNVRNVPLPGDAAAGHPDRASGHPKPHSRSSSAYRKSASTHHKSVFSHSKAASGHLKPVSGHSKPASGHPKSATVYPKPASVHFKADSVHFKGDSVHFKPDSVHFKPASSNPKPITGHHVSAGSHSKSAFSAATSHPKPTTGHIKPATSHAEPTTADYPKPATTSHPKPTAADNPELSASHCPEIPAIAHPVSDDSDLPESASSPAAGPTKPAASQLESDTIADLPDPTVVTTSTNDYHDVVVVDVEDDPDEMAV

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service