Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401404

Sigma-Aldrich

Anti-WNT9B antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

WNT14B, WNT15

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WNT9B(7484)

General description

The WNT gene family consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region. (provided by RefSeq)

Immunogen

WNT9B (AAH64534.1, 1 a.a. ~ 329 a.a) full-length human protein.

Sequence
MRPPPALALAGLCLLALPAAAASYFGLTGREVLTPFPGLGTAAAPAQGGAHLKQCDLLKLSRRQKQLCRREPGLAETLRDAAHLGLLECQFQFRHERWNCSLEGRTGLLKRGFKETAFLYAVSSAALTHTLARACSAGRMERCTCDDSPGLESRQAWQWGVCGDNLKYSTKFLSNFLGSKRGNKDLRARADAHNTHVGIKAVKSGLRTTCKCHGVSGSCAVRTCWKQLSPFRETGQVLKLRYDSAVKVSSATNEALGRLELWAPARQGSLTKGLAPRSGDLVYMEDSPSFCRPSKYSPGTAGWSAVARSQLITTSTSRIQAILPSQLPE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service