Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1401320

Sigma-Aldrich

Anti-PTH antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

PTH1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... PTH(5741)

General description

The protein encoded by this gene is a hormone secreted by parathyroid cells. This hormone elevates blood Ca2+ level by dissolving the salts in bone and preventing their renal excretion. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH). (provided by RefSeq)

Immunogen

PTH (NP_000306.1, 1 a.a. ~ 115 a.a) full-length human protein.

Sequence
MIPAKDMAKVMIVMLAICFLTKSDGKSVKKRSVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFVALGAPLAPRDAGSQRPRKKEDNVLVESHEKSLGEADKADVNVLTKAKSQ

Biochem/physiol Actions

PTH (parathyroid hormone) is an important regulatory hormone for calcium homeostasis. In chronic kidney disease, hyperparathyroidism caused due to increased levels of PTH leads to arteriosclerotic fibrosis and calcification. Elevation in the levels of PTH might be associated with hypertension, bone disease and also in few cases it results in death due to cardiovascular disease. PTH is known to induce the secretion of aldosterone hormone in adrenocortex.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PTH Is a Promising Auxiliary Index for the Clinical Diagnosis of Aldosterone-Producing Adenoma.
Zhang LX
American Journal of Hypertension, 29(5), 575-581 (2016)
Genetic Variants Associated with Circulating Parathyroid Hormone.
Robinson-Cohen C
Journal of the American Society of Nephrology, 28(5), 1553-1565 (2017)
Elevated PTH induces endothelial-to-chondrogenic transition in aortic endothelial cells.
Wu M
American Journal of Physiology: Renal Physiology, 312(3), 346-344 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service