SAB1400856
Anti-FLJ38288 antibody produced in mouse
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-DKFZp686K10134
Select a Size
$541.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$541.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
indirect immunofluorescence: suitable
western blot: 1 μg/mL
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... FLJ38288(284309)
Immunogen
Sequence
MLENLTLISSLGCWYGAKDETPSKQTLSIQQESPLRTHWTGVCTKKVHLWGMCGPLLGDILHQGTQHNQKLNGFGAYEKKLDDDANHHQDQKQHIGEKSYRSNAKGTSFVKNCKFHMSHEPFIFHEVGKDFLSSLRLLQQEDIHTSGKSNFETKHGIPLQGGKTHYICGESTIPFSNKHSLVLHQRLLPREGPYVCSDSGKFTSKSNSFNNHQGVRTGKRPYQCGQCDESFWYKAHLTEHQRVHTGERPYECGECDKSFSHKHSLVDHQRVHTGERPYECDECGKSFSHKRSLVHHQRVHTGERPYQCGECGKSFNHKCNLIQHQRVHTGERPFECTACGKLFRSNSHLKEHQRVHTGERPYECKECRKSFRYKSHLTEHQRVHTGERPYECRECGKCFHQKGSLIQHQQIHSGERPHECGECGKCFHQKGSLIRHQQIHSGERPHECGECGKCFRQKGNLIKHQRVHTGERHHEC
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service