Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400843

Sigma-Aldrich

Anti-NALP11 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-NOD17, Anti-PAN10, Anti-PYPAF6

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NALP11(204801)

Related Categories

General description

NALPs are cytoplasmic proteins that form a subfamily within the larger CATERPILLER protein family. Most short NALPs, such as NALP11, have an N-terminal pyrin (MEFV; MIM 608107) domain (PYD), followed by a NACHT domain, a NACHT-associated domain (NAD), and a C-terminal leucine-rich repeat (LRR) region. The long NALP, NALP1 (MIM 606636), also has a C-terminal extension containing a function to find domain (FIIND) and a caspase recruitment domain (CARD). NALPs are implicated in the activation of proinflammatory caspases (e.g., CASP1; MIM 147678) via their involvement in multiprotein complexes called inflammasomes (Tschopp et al., 2003 [PubMed 12563287]).[supplied by OMIM

Immunogen

NLRP11 (NP_659444.1, 1 a.a. ~ 1033 a.a) full-length human protein.

Sequence
MAESDSTDFDLLWYLENLSDKEFQSFKKYLARKILDFKLPQFPLIQMTKEELANVLPISYEGQYIWNMLFSIFSMMRKEDLCRKIIGRRNRNQEACKAVMRRKFMLQWESHTFGKFHYKFFRDVSSDVFYILQLAYDSTSYYSANNLNVFLMGERASGKTIVINLAVLRWIKGEMWQNMISYVVHLTSHEINQMTNSSLAELIAKDWPDGQAPIADILSDPKKLLFILEDLDNIRFELNVNESALCSNSTQKVPIPVLLVSLLKRKMAPGCWFLISSRPTRGNNVKTFLKEVDCCTTLQLSNGKREIYFNSFFKDRQRASAALQLVHEDEILVGLCRVAILCWITCTVLKRQMDKGRDFQLCCQTPTDLHAHFLADALTSEAGLTANQYHLGLLKRLCLLAAGGLFLSTLNFSGEDLRCVGFTEADVSVLQAANILLPSNTHKDRYKFIHLNVQEFCTAIAFLMAVPNYLIPSGSREYKEKREQYSDFNQVFTFIFGLLNANRRKILETSFGYQLPMVDSFKWYSVGYMKHLDRDPEKLTHHMPLFYCLYENREEEFVKTIVDALMEVTVYLQSDKDMMVSLYCLDYCCHLRTLKLSVQRIFQNKEPLIRPTASQMKSLVYWREICSLFYTMESLRELHIFDNDLNGISERILSKALEHSSCKLRTLKLSYVSTASGFEDLLKALARNRSLTYLSINCTSISLNMFSLLHDILHEPTCQISHLSLMKCDLRASECEEIASLLISGGSLRKLTLSSNPLRSDGMNILCDALLHPNCTLISLVLVFCCLTENCCSALGRVLLFSPTLRQLDLCVNRLKNYGVLHVTFPLLFPTCQLEELHLSGCFFSSDICQYIAIVIATNEKLRSLEIGSNKIEDAGMQLLCGGLRHPNCMLVNIGLEECMLTSACCRSLASVLTTNKTLERLNLLQNHLGNDGVAKLLESLISPDCVLKVVGLPLTGLNTQTQQLLMTVKERKPSLIFLSETWSLKEGREIGVTPASQPGSIIPNSNLDYMFFKFPRMSAAMRTSNTASRQPL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service