Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400734

Sigma-Aldrich

Anti-RIP antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-HRIP, Anti-MGC4189

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RIP(84268)

Immunogen

RPAIN (NP_001028174.1, 1 a.a. ~ 219 a.a) full-length human protein.

Sequence
MAESLRSPRRSLYKLVGSPPWKEAFRQRCLERMRNSRDRLLNRYRQAGSSGPGNSQNSFLVQEVMEEEWNALQSVENCPEDLAQLEELIDMAVLEEIQQELIKQEQSIISEYEKSLQFDEKCLSIMLAEWEANPLICPVCTKYNLRITSGVVVCQCGLSIPSHSSELTEQKLRACLEGSINEHSAHCPHTPEFSVTGGTEEKSSLLMSCLACDTWAVIL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caiguo Zhang et al.
Molecular oncology, 12(4), 476-494 (2018-01-30)
Cullin 4B, a member of the Cullins, which serve as scaffolds to facilitate the assembly of E3 ligase complexes, is aberrantly expressed in many cancers, including osteosarcoma. Recently, we observed that CUL4B forms the CRL4BDCAF11 E3 ligase, which specifically ubiquitinates

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service