Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1400731

Sigma-Aldrich

Anti-GTF2IRD2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FLJ21423, Anti-GTF2IRD2A

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene is unusual in that its coding sequence is mostly derived from Charlie8 repeat elements. However, there is mRNA and EST evidence to suggest that this gene is transcribed, and the encoded protein has a homolog in mouse, with which it shares 78% sequence identity. The exact function of this gene product is not known. It is inferred to be a transcription factor based on the presence of GTF2I-like repeats (containing helix-loop-helix motifs), also found in other proteins such as GTF2IRD1 and GTF2I. These three genes are clustered together on chr 7q11.23. (provided by RefSeq)

Immunogen

GTF2IRD2 (AAH67859, 1 a.a. ~ 498 a.a) full-length human protein.

Sequence
MAQVAVSTLPVEEESSSETRMVVTFLVSALESMCKELAKSKAEVACIAVYETDVFVVGTERGCAFVNARTDFQKDFAKYCVAEGLCEVKPPCPVNGMQVHSGETEILRKAVEDYFCFCYGKALGTTVMVPVPYEKMLRDQSAVVVQGLPEGVAFQHPENYDLATLKWILENKAGISFIINRPFLGPESQLGGPGMVTDAERSIVSPSESCGPINVKTEPMEDSGISLKAEAVSVKKESEDPNYYQYNMQGSHPSSTSNEVIEMELPMEDSTPLVPSEEPNEDPEAEVKIEGNTNSSSVTNSAAGVEDLNIVQVTVPDNEKERLSSIEKIKQLREQVNDLFSRKFGEAIGVDFPVKVPYRKITFNPGCVVIDGMPPGVVFKAPGYLEISSMRRILEAAEFIKFTVIRPLPGLELSNGLLEAGAGGLLEAGSLRPAWATWQDPISTKNLKISWKQWLTPVAPATWQAEAGGSLELRSLRQQRTIIAILHSSLGDRTRPCL

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service