Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1400692

Sigma-Aldrich

Anti-TSEN34 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-LENG5, Anti-SEN34, Anti-SEN34L

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TSEN34(79042)

Related Categories

General description

tRNA splicing is a fundamental process required for cell growth and division. SEN34 is a subunit of the tRNA splicing endonuclease, which catalyzes the removal of introns, the first step in tRNA splicing (Paushkin et al., 2004 [PubMed 15109492]).[supplied by OMIM

Immunogen

TSEN34 (AAH00944.1, 1 a.a. ~ 390 a.a) full-length human protein.

Sequence
MLVVEVANGRSLVWGAEAVQALRERLGVGGRTVGALPRGPRQNSRLGLPLLLMPEEARLLAEIGAVTLVSAPRPDSRHHSLALTSFKRQQEESFQEQSALAAEARETRRQEVLEKITEGQAAKKQKLEQASGASSSQEAGSSQAAKEDETSDGQASGEQEEAGPSSSQAGPSNGVAPLPRSALLVQLATARPRPVKARPLDWRVQSKDWPHAGRPAHELRYSIYRDLWERGFFLSAAGKFGGDFLVYPGDPLRFHAHYIAQCWAPEDTSHSKTWLLLGALEPASERPCSSVLRSLMVRWSTPPCNGPACSELQRPRGCGCVGSKSLSGCSPALLWESRTSSYLSPRLVFDSRFSNTTSFLCSSLFQSTYWLCFCSYLFSHCELPENGAWV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service