Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400663

Sigma-Aldrich

Anti-MRPS35 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DKFZp762P093, Anti-HDCMD11P, Anti-MDS023, Anti-MGC104278, Anti-MRP-S28, Anti-MRPS28

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MRPS35(60488)

General description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that has had confusing nomenclature in the literature. Pseudogenes corresponding to this gene are found on chromosomes 3p, 5q, and 10q. (provided by RefSeq)

Immunogen

MRPS35 (NP_068593.2, 1 a.a. ~ 323 a.a) full-length human protein.

Sequence
MAAAALPAWLSLQSRARTLRAFSTAVYSATPVPTPSLPERTPGNERPPRRKALPPRTEKMAVDQDWPSVYPVAAPFKPSAVPLPVRMGYPVKKGVPMAKEGNLELLKIPNFLHLTPVAIKKHCEALKDFCTEWPAALDSDEKCEKHFPIEIDSTDYVSSGPSVRNPRARVVVLRVKLSSLNLDDHAKKKLIKLVGERYCKTTDVLTIKTDRCPLRRQNYDYAVYLLTVLYHESWNTEEWEKSKTEADMEEYIWENSSSERNILETLLQMKAAEKNMEINKEELLGTKEIEEYKKSVVSLKNEEENENSISQYKESVKRLLNVT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service