Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1400615

Sigma-Aldrich

Anti-IMPACT antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MGC33718

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$541.00

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
50 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IMPACT(55364)

Related Categories

Immunogen

IMPACT (AAH34016, 1 a.a. ~ 320 a.a) full-length human protein.

Sequence
MAEGDAGSDQRQNEEIEAMAAIYGEEWCVIDDCAKIFCIRISDDIDDPKWTLCLQVMLPNEYPGTAPPIYQLNAPWLKGQERADLSNSLEEIYIQNIGESILYLWVEKIRDVLIQKSQMTEPGPDVKKKTEEEDVECEDDLILACQPESSVKALDFDISETRTEVEVEELPPIDHGIPITDRRSTFQAHLAPVVCPKQVKMVLSKLYENKKIASATHNIYAYRIYCEDKQTFLQDCEDDGETAAGGRLLHLMEILNVKNVMVVVSRWYGGILLGPDRFKHINNCARNILVEKNYTNSPEESSKALGKNKKVRKDKKRNEH

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service