SAB1400431
Anti-MTMR15 antibody produced in mouse
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-DKFZp451H236, Anti-DKFZp686K16147, Anti-KIAA1018
Select a Size
$541.00
Select a Size
About This Item
$541.00
biological source
mouse
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
immunofluorescence: suitable
western blot: 1 μg/mL
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MTMR15(22909)
General description
Immunogen
Sequence
MSEGKPPDKKKRPRRSLSISKNKKKASNSIISCFNNAPPAKLACPVCSKMVPRYDLNRHLDEMCANNDFVQVDPGQVGLINSNVSMVDLTSVTLEDVTPKKSPPPKTNLTPGQSDSAKREVKQKISPYFKSNDVVCKNQDELRNRSVKVICLGSLASKLSRKYVKAKKSIDKDEEFAGSSPQSSKSTVVKSLIDNSSEIEDEDQILENSSQKENVFKCDSLKEECIPEHMVRESKIMEAESQKATRECEKSALTPGFSDNAIMLFSPDFTLRNTLKSTSEDSLVKQECIKEVVEKREACHCEEVKMTVASEAKIQLSDSEAKSHSSADDASAWSNIQEAPLQDDSCLNNDIPHSIPLEQGSSCNGPGQTTGHPYYLRSFLVVLKTVLENEDDMLLFDEQEKGIVTKFYQLSATGQKLYVRLFQRKLSWIKMTKLEYEEIALDLTPVIEELTNAGFLQTESELQELSEVLELLSAPELKSLAKTFHLVNPNGQKQQLVDAFLKLAKQRSVCTWGKNKPGIGAVILKRFCWLLLQ
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service