Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB1400212

Sigma-Aldrich

Anti-PLCG2 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

proximity ligation assay: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PLCG2(5336)

General description

Enzymes of the phospholipase C family catalyze the hydrolysis of phospholipids to yield diacylglycerols and water-soluble phosphorylated derivatives of the lipid head groups. A number of these enzymes have specificity for phosphoinositides. Of the phosphoinositide-specific phospholipase C enzymes, C-beta is regulated by heterotrimeric G protein-coupled receptors, while the closely related C-gamma-1 (PLCG1; MIM 172420) and C-gamma-2 enzymes are controlled by receptor tyrosine kinases. The C-gamma-1 and C-gamma-2 enzymes are composed of phospholipase domains that flank regions of homology to noncatalytic domains of the SRC oncogene product, SH2 and SH3.[supplied by OMIM

Immunogen

PLCG2 (AAH07565.1, 1 a.a. ~ 1265 a.a) full-length human protein.

Sequence
MSTTVNVDSLAEYEKSQIKRALELGTVMTVFSFRKSTPERRTVQVIMETRQVAWSKTADKIEGFLDIMEIKEIRPGKNSKDFERAKAVRQKEDCCFTILYGTQFVLSTLSLAADSKEDAVNWLSGLKILHQEAMNASTPTIIESWLRKQIYSVDQTRRNSISLRELKTILPLINFKVSSAKFLKDKFVEIGAHKDELSFEQFHLFYKKLMFEQQKSILDEFKKDSSVFILGNTDRPDASAVYLRDFQRFLIHEQQEHWAQDLNKVRERMTKFIDDTMRETAEPFLFVDEFLTYLFSRENSIWDEKYDAVDMQDMNNPLSHYWISSSHNTYLTGDQLRSESSPEAYIRCLRMGCRCIELDCWDGPDGKPVIYHGWTRTTKIKFDDVVQAIKDHAFVTSSFPVILSIEEHCSVEQQRHMAKAFKEVFGDLLLTKPTEASADQLPSPSQLREKIIIKHKKLGPRGDVDVNMEDKKDEHKQQGELYMWDSIDQKWTRHYCAIADAKLSFSDDIEQTMEEEVPQDIPPTELHFGEKWFHKKVEKRTSAEKLLQEYCMETGGKDGTFLVRESETFPNDYTLSFWRSGRVQHCRIRSTMEGGTLKYYLTDNLTFSSIYALIQHYRETHLRCAEFELRLTDPVPNPNPHESKPWYYDSLSRGEAEDMLMRIPRDGAFLIRKREGSDSYAITFRARGKVKHCRINRDGRHFVLGTSAYFESLVELVSYYEKHSLYRKMRLRYPVTPELLERYNMERDINSLYDVSRMYVDPSEINPSMPQRTVKALYDYKAKRSDELSFCRGALIHNVSKEPGGWWKGDYGTRIQQYFPSNYVEDISTADFEELEKQIIEDNPLGSLCRGILDLNTYNVVKAPQGKNQKSFVFILEPKQQGYPPVEFATDRVEELFEWFQSIREITWKIDTKENNMKYWEKNQSIAIELSDLVVYCKPTSKTKDNLENPDFREIRSFVETKADSIIRQKPVDLLKYNQKGLTRVYPKGQRVDSSNYDPFRLWLCGSQMVALNFQTADKYMQMNHALFSLNGRTGYVLQPESMRTEKYDPMPPESQRKILMTLTVKVLGARHLPKLGRSIACPFVEVEICGAEYDNNKFKTTVVNDNGLSPIWAPTQEKVTFEIYDPNLAFLRFVVYEEDMFSDPNFLAHATYPIKAVKSGFRSVPLKNGYSEDIELASLLVFCEMRPVLESEEELYSSCRQLRRRQEELNNQLFLYDTHQNLRNANRDALVKEFSVNENQLQLYQEKCNKRLREKRVSNSKFYS

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service