Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1307183

Sigma-Aldrich

ANTI-SHCBP1(C-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

SHC SH2 domain-binding protein 1, SHCBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

75690 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SHCBP1(79801)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Houfa Geng et al.
Frontiers in endocrinology, 11, 613879-613879 (2021-03-16)
Papillary thyroid carcinoma (PTC) is the most common thyroid cancer with a rapidly increasing incidence globally. Bioinformatics analyses suggested that SHCBP1 (SHC SH2 Domain-Binding Protein 1) was significantly up-regulated in PTC tumor tissues, which was further confirmed by immunohistochemical staining
Chunhong Ren et al.
Applied immunohistochemistry & molecular morphology : AIMM, 29(2), 136-143 (2020-08-10)
Esophageal cancer (EC) is known as a type of common malignant tumor, with the incidence ranking eighth worldwide. Because of the high metastasis of advanced EC, the total survival rate has been quite low. Esophageal squamous cell carcinoma (ESCC) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service