Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1306765

Sigma-Aldrich

ANTI-MAP3K12 antibody produced in mouse

crude ascites, buffered aqueous solution

Synonym(s):

Dual leucine zipper bearing kinase, Leucine-zipper protein kinase, MAP3K12, Mitogen-activated protein kinase kinase kinase 12, ZPK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

crude ascites

antibody product type

primary antibodies

clone

monoclonal

form

buffered aqueous solution

mol wt

93219 Da

species reactivity

human, mouse

technique(s)

western blot: 1:800

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAP3K12(7786)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service