Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1306696

Sigma-Aldrich

ANTI-CB2(C-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

CNR2, CX5, Cannabinoid receptor 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

39681 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CNR2(1269)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hai-Ying Zhang et al.
Acta pharmacologica Sinica, 40(3), 398-409 (2018-07-04)
Cannabinoid CB1 receptors are highly expressed in the brain and functionally modulate presynaptic neurotransmitter release, while cannabinoid CB2 receptors (CB2Rs) were initially identified in the spleen and regarded as peripheral cannabinoid receptors. Recently, growing evidence indicates the presence of functional
Michał Biernacki et al.
Advances in medical sciences, 64(1), 15-23 (2018-09-23)
The effect of chronic administration of [3-(3-carbamoylphenyl)phenyl] N-cyclohexylcarbamate (URB597), inhibitor of fatty acid amide hydrolase (FAAH) that hydrolyzes anandamide, on cross-talk between endocannabinoid system, oxidative status and pro-inflammatory factors in the liver of spontaneously hypertensive rats (SHRs) was investigated. Experiments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service