Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1303156

Sigma-Aldrich

ANTI-AP1S1 (N-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

AP-1 complex subunit sigma-1A, AP19, AP1S1, Adapter-related protein complex 1 sigma-1A subunit, Adaptor protein complex AP-1 sigma-1A subunit, CLAPS1, Clathrin assembly protein complex 1 sigma-1A small chain, Clathrin coat assembly protein AP19, Golgi adaptor HA1/AP1 adaptin sigma-1A subunit, HA1 19 kDa subunit, Sigma 1a subunit of AP-1 clathrin, Sigma-adaptin 1A, Sigma1A-adaptin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

18733 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AP1S1(1174)

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service