Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300984

Sigma-Aldrich

Anti-EEF2K (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-EEF-2 kinase, Anti-EEF-2K, Anti-EEF2K, Anti-Elongation factor 2 kinase, Calcium/calmodulin-dependent eukaryotic elongation factor 2 kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEF2K(29904)

General description

Eukaryotic elongation factor 2 kinase (eEF2k) phosphorylates and inactivates eEF2, thereby inhibiting peptide-chain elongation. eEF2k, which is Ca2+ and calmodulin dependent, can be activated by PKA in response to stress-induced elevation of cAMP levels. eEF2k expression is also modulated by a wide range of stimuli that promote cell growth and protein synthesis. Phosphorylation of eEF2k by p90RSK and p70 S6 kinase at Ser366 or by SAPK4/p38d at Ser359, inactivates eEF2k, which facilitates the dephosphorylation of eEF2, and thus promotes translation.

Immunogen

EEF2K (NP_037434, 605-640)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human elongation factor 2 kinase.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service