Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300978

Sigma-Aldrich

Anti-DGKI (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DGKI(9162)

Immunogen

DGKI (139-174)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human DGKI.

Application

Anti-DGKI (N-term) antibody produced in rabbit is suitable for immunohistochemistry at a dilution of 1:50-1:100 and indirect ELISA at a dilution of 1:1000.

Biochem/physiol Actions

DGKI (Diacylglycerol kinase iota) is mainly involved in the maintenance of intracellular concentration of diacylglycerol by phosphorylating other lipids in addition to DAG. DGKI has been predicted to be a potential gene for tumor suppression in colorectal cancers and mutation in DGKI has been linked to the melanoma.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of Chromosomal Instability and Epigenetics in Colorectal Cancers Lacking ??-Catenin/TCF Regulated Transcription
Gastroenterology Research and Practice (2016)
The Role of Chromosomal Instability and Epigenetics in Colorectal Cancers Lacking ??-Catenin/TCF Regulated Transcription
Wael M. Abdel-Rahman, et al.
Gastroenterology Research and Practice (2016)
Matthew K Topham et al.
Biochimica et biophysica acta, 1790(6), 416-424 (2009-04-15)
The mammalian diacylglycerol kinases (DGK) are a group of enzymes having important roles in regulating many biological processes. Both the product and the substrate of these enzymes, i.e. diacylglycerol and phosphatidic acid, are important lipid signalling molecules. Each DGK isoform

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service