Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300915

Sigma-Aldrich

Anti-PRKCDBP (center) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-HSRBC, Anti-PRKCDBP, Anti-Protein kinase C δ-binding protein, Anti-SRBC, Serum deprivation response factor-related gene product that binds to C-kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

PRKCDBP was identified as a binding protein of the protein kinase C, delta (PRKCD). The expression of this protein in cultured cell lines is strongly induced by serum starvation, and was found to be down-regulated in various cancer cell lines, suggesting the possible tumor suppressor function of this protein.

Immunogen

PRKCDBP (NP_659477, 118-154)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human PRKCDBP.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service