Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300784

Sigma-Aldrich

Anti-Mouse Kif5C-1 (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-Kif5c, Anti-Kinesin heavy chain isoform 5C, Anti-Kinesin heavy chain neuron-specific 2, Anti-Nkhc2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

General description

Kinesins are microtubule-based motor proteins involved in the transport of organelles in eukaryotic cells. They typically consist of 2 identical, approximately 110- to 120-kD heavy chains and 2 identical, approximately 60- to 70-kD light chains. The heavy chain contains 3 domains: a globular N-terminal motor domain, which converts the chemical energy of ATP into a motile force along microtubules in 1 fixed direction; a central alpha-helical rod domain, which enables the 2 heavy chains to dimerize; and a globular C-terminal domain, which interacts with light chains and possibly an organelle receptor.

Immunogen

KIF5C (NP_032475, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse Kif5C-1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service