Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300783

Sigma-Aldrich

Anti-CLC4 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CLCN4, Anti-Chloride channel protein 4, Anti-ClC-4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
indirect immunofluorescence: 1:10-1:50
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLC4(1183)

General description

CLCN4 (chloride channel 4) gene codes for the chloride/hydrogen ion exchanger CIC-4. It is abundantly expressed in brain. ClC-4 belongs to the ClC transporter branch. It is present in the intracellular membranes of the brain, muscle and liver. This gene is located on human chromosome Xp22.3.

Immunogen

CLC4 (NP_001821, 662-699)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human CLC4.

Biochem/physiol Actions

CLCN4 (chloride channel 4) is associated with X-linked intellectual disability and epilepsy. ClC-4 helps to interchange anion/proton with variable stoichiometry.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Channel-like slippage modes in the human anion/proton exchanger ClC-4.
Alekov AK and Fahlke C
The Journal of General Physiology, 133(5), 485-496 (2009)
De novo and inherited mutations in the X-linked gene CLCN4 are associated with syndromic intellectual disability and behavior and seizure disorders in males and females.
Palmer EE, et al.
Molecular Psychiatry, 23(2), 222?230-222?230 (2016)
Application of microarrays to the analysis of the inactivation status of human X-linked genes expressed in lymphocytes.
Craig IW, et al.
European Journal of Human Genetics, 12(8), 639-639 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service