Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300771

Sigma-Aldrich

Anti-DSCR1L1 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Calcipressin-2, Anti-MCIP2, Anti-Thyroid hormone-responsive protein ZAKI-4, Down syndrome candidate region 1-like 1, Myocyte-enriched calcineurin interacting protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DSCR1L1(10231)

Related Categories

General description

DSCR1L1 inhibits calcineurin-dependent transcriptional responses by binding to the catalytic domain of calcineurin A. This protein may play a role during central nervous system development. Expression is detected in fibroblasts, heart, brain, liver, and skeletal muscle but not in placenta, lung, kidney and pancreas. Expression of both transcripts is upregulated by physiologic concentrations of the thyroid hormone triiodothyroxine.

Immunogen

DSCR1L1 (NP_005813, 71-106)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human DSCR1L1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service