Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300739

Sigma-Aldrich

Anti-BIRC6 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BIRC6(57448)

General description

BIRC6 is a member of a family of proteins that inhibit apoptosis, likely through interference with activation of ICE-like proteases. BIRC6, which contains a single BIR domain and a ubiquitin-conjugating enzyme domain, is expressed in four of six brain cancers (gliomas), and one of five ovarian cancers in a variety of human cancer cell lines. Brain cancer cell lines that overexpress BIRC6 demonstrate multi-drug resistance BIRC6 may protect cancer cells from undergoing apoptosis and participate in tumorigenesis and drug resistance.

Immunogen

BIRC6 (BIR6_HUMAN, 27-62)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human BIRC6.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service