Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300731

Sigma-Aldrich

Anti-Phospho-Leo1-pS551 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-RDL, Anti-RNA polymerase-associated protein LEO1, Replicative senescence down-regulated leo1-like protein

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
$437.00
100 μG
$455.00

$437.00


Check Cart for Availability


Select a Size

Change View
400 μL
$437.00
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$437.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

dot blot: suitable
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer551)

Gene Information

human ... LEO1(123169)

General description

The PAF1 complex is a multifunctional complex. The PAF1 complex interacts with POLR2A. It may be involved in both initiation and elongation, histone methylation and RNA processing. Overexpression of LEO1 induces cell growth arrest and premature senescence of fibroblasts.

Immunogen

LEO1 (NP_620147, 534-567)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S551 of human LEO1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service