Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300726

Sigma-Aldrich

Anti-Phospho-SLAMF1-pY281 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD150 antigen, Anti-CDw150, Anti-IPO-3, Anti-SLAMF1, Anti-Signaling lymphocytic activation molecule [Precursor]

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
$437.00
100 μG
$455.00

$437.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
400 μL
$437.00
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$437.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

dot blot: suitable
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr281)

Gene Information

human ... SLAMF1(6504)

General description

SLAMF1 is a high-affinity self-ligand important in bidirectional T-cell to B-cell stimulation. SLAM-induced signal-transduction events in T-lymphocytes are different from those in B-cells. Two modes of SLAM signaling are likely to exist: one in which the inhibitor SH2D1A acts as a negative regulator and another in which protein-tyrosine phosphatase 2C (PTPN11)-dependent signal transduction operates.

Immunogen

SLAMF1 (NP_003028, 264-296)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding Y281 of human SLAMF1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service