Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300657

Sigma-Aldrich

Anti-TPST1 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-TPST-1, Anti-Tyrosylprotein sulfotransferase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TPST1(8460)

General description

The importance of tyrosine O-sulfation in protein-protein interactions is illustrated by the sulfation of tyrosine residues in the leukocyte adhesion molecule P-selectin glycoprotein ligand-1; sulfation is required for its binding to P-selectin on activated endothelium. This enzyme represents a new class of Golgi sulfotransferases that catalyze tyrosine O-sulfation of many protein substrates involved in diverse physiologic functions including inflammation, hemostasis, body weight and reproductive physiology.

Immunogen

TPST1 (NP_003587, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human TPST1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service