Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300607

Sigma-Aldrich

Anti-RPC5 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DNA-directed RNA polymerases III 80 kDa polypeptide, Anti-KIAA1452, Anti-POLR3E, Anti-RNA polymerase III subunit 5, Anti-RPC5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPC5(55718)

General description

RNA polymerase III synthesizes RNA components of the protein synthesis, pre-mRNA splicing, and tRNA processing apparatuses. The holoenzyme consists of about 15 different subunits. The RPC5 subunit is essential for efficient transcription from both the type 2 VAI and type 3 U6 RNA polymerase III promoters.

Immunogen

RPC5 (NP_060589, 239-273)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of human RPC5.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service