Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300538

Sigma-Aldrich

Anti-PAT2 (Slc36a2) (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-PAT2, Anti-Proton-coupled amino acid transporter 2, Anti-Proton/amino acid transporter 2, Anti-Solute carrier family 36 member 2, Anti-Tramdorin-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAT2(246235)

General description

PAT2 is involved in a pH-dependent electrogenic neuronal transport and sequestration of small amino acids amino acids such as glycine, alanine and proline. It is inhibited by sarcosine.

Immunogen

PAT2 (AAI66414.1, 421-456)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of rat PAT2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service