Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB1300531

Sigma-Aldrich

Anti-HSP40 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSP40(3337)

General description

DnaJ (Hsp40) belongs to the DnaJ-class of molecular chaperones with a C-terminal Zn finger domain. HSP40 (DnaJ) together with DnaK and GrpE form a molecular chaperone that is involved in formation of protein complexes, protein folding, prevention of protein aggregation, and protein turnover and export. Several human neurodegenerative diseases involve the expansion of a polyglutamine within the disease proteins. Molecular chaperones such as HSP40 complexes can modulate polyglutamine pathogenesis In transgenic Drosophila disease models of Machado-Joseph disease and Huntington disease Hdj1, the Drosophila homolog to human HSP40, demonstrates substrate specificity for polyglutamine proteins suppression in combination with other molecular chapterones of neurotoxicity, and altered solubility of mutant polyglutamine proteins.

Immunogen

HSP40 (DJB1_HUMAN, Full length)
This antibody is generated from rabbits immunized with a recombinant protein encoding full length of human HSP40.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service