Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300520

Sigma-Aldrich

Anti-ELP3 (N-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-EC 2.3.1.48, Anti-ELP3, Anti-Elongator complex protein 3, Anti-HELP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELP3(55140)

General description

The gene ELP3 (elongator complex protein 3) is mapped to human chromosome 8p21.1. The encoded protein has a HAT (histone acetyltransferase) domain.

Immunogen

ELP3 (NP_060561, 14-54)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human ELP3.

Biochem/physiol Actions

ELP3 (elongator complex protein 3) is a catalytic subunit of the histone acetyltransferase elongator complex. It enhances histone H3 acetylation, thereby regulating gene regulation in neurons. Dysregulation of ELP3 activity causes neuronal disorders, such as familial dysautonomia and amyotrophic lateral sclerosis. It might be a potential acetyltransferase for glucose-6-phosphate dehydrogenase. The ELP3 gene promoter is hypermethylated in invasive ductal breast carcinoma, causing transcriptional inactivation of the gene.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constitutional trisomy 8 mosaicism as a model for epigenetic studies of aneuploidy.
Davidsson J, et al.
Epigenetics & Chromatin, 6, 18-18 (2013)
hElp3 directly modulates the expression of HSP70 gene in HeLa cells via HAT activity.
Li F, et al.
PLoS ONE, 6, e29303-e29303 (2011)
Regulation of G6PD acetylation by SIRT2 and KAT9 modulates NADPH homeostasis and cell survival during oxidative stress.
Wang YP, et al.
The Embo Journal, 33, 1304-1320 (2014)
The histone acetyltransferase Elp3 plays in active role in the control of synaptic bouton expansion and sleep in Drosophila.
Singh N, et al.
Journal of Neurochemistry, 115, 493-504 (2010)
Promoter hypermethylation may be an important mechanism of the transcriptional inactivation of ARRDC3, GATA5, and ELP3 in invasive ductal breast carcinoma.
Wang D
Molecular and Cellular Biochemistry, 396, 67-77 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service