Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300503

Sigma-Aldrich

Monoclonal Anti-mobC antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Mobc, Anti-Mobilization protein C

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

19CT.2.8.14, monoclonal

form

buffered aqueous solution

species reactivity

Neisseria gonorrhoeae

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

The mobilization region of the 7.4 kb beta-lactamase plasmid pSJ7.4 from Neisseria gonorrhoeae was characterized. The 3.2 kb HindIII-BamHI fragment of pSJ7.4 was mobilized between Escherichia coli strains by conjugative plasmid RK2. Selected restriction enzyme-generated deletions of this fragment were subcloned in pACYC177 to obtain constructs that were suitable for analysis of the mobilization region. Mating experiments showed that a highly conserved 1.9 kb DNA region within coordinates 4096-5997 is required for mobilization of pSJ7.4. This region contains two genes encoding the mobilization protein MobA, another protein, MobC, putatively involved in mobilization from some hosts, and an intergenic oriT. The 168 bp intergenic sequence also contains the promoters for mobA and mobC in an arrangement that suggests divergent transcription and autoregulation from oriT. The 56 kDa MobA was expressed in E. coli as a (6x)His-Tag fusion protein. Purified MobA specifically induced plasmid relaxation by nicking at the oriT. MobA is exceptional because the N-terminal region alone can mobilize pSJ7.4, albeit at a lower frequency than the full-length protein, even in the absence of MobC. The carboxyl terminal region of MobA did not share homology with other mobilization proteins, but may be involved in promoting efficient transfer of pSJ7.4.

Immunogen

MOBC (AAO45531, )
This antibody was raised using purified His-tagged recombinant full length Neisseria gonorrhoeae plasmid pSJ7.4 mobilization protein C (mobC)and mobilization protein A (mobA).

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service