Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300502

Sigma-Aldrich

Monoclonal Anti-H1L antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Late protein H1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

20CT26.44.10.3 / 20CT26.44.10.14, monoclonal

form

buffered aqueous solution

species reactivity

vaccinia virus

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

This protein shows activity toward tyrosine-protein phosphate as well as with serine-protein phosphate. Purified recombinant H1L fusion protein was used to produced this monoclonal antibody.

Immunogen

H1L (N/A)
Purified recombinant H1L fusion protein was used to produced this monoclonal antibody.

Physical form

Purified monoclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service