Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300494

Sigma-Aldrich

Anti-PTP-U (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTP-U(10076)

General description

PTP-U, a member of the receptor class 2B subfamily of protein-tyrosine phosophatases, is involved in regulation of processes involving cell contact and adhesion such as growth control, tumor invasion, and metastasis. It forms complexes with beta-catenin and gamma-catenin/plakoglobin. This Type I membrane protein is found at high levels in brain, pancreas, and skeletal muscle; less in colon, kidney, liver, stomach, and uterus; no expression is observed in placenta and spleen. PTP-U is up-regulated upon cell contact. The protein contains 4 fibronectin type III domains, 1 immunoglobulin-like C2-type domain, 1 MAM domain, and 2 protein-tyrosine phosphatase domains.

Immunogen

PTP-U (Q92729, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human PTP-U.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service