Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300450

Sigma-Aldrich

Anti-Haspin GSG2 (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-GSG2, Anti-H-Haspin, Anti-Serine/threonine-protein kinase Haspin, Germ cell-specific gene 2 protein, Haploid germ cell-specific nuclear protein kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GSG2(83903)

Related Categories

General description

Post-translational modifications of conserved N-terminal tail residues in histones regulate many aspects of chromosome activity. Mitotic phosphorylation of H3 Thr 3 occurs in prophase and dephosphorylation during anaphase. Haspin, a dual serine/threonine kinase, plays an important role in regulation of chromosome and spindle function during mitosis and meiosis via its function in phosphorylation of the threonine residue in the third position of histone 3 (Thr3).

Immunogen

HASPIN GSG2 (NP_114171, 320-355)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the central region of human haspin.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service