Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300438

Sigma-Aldrich

Anti-PI3KR5 (center) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-P101-PI3K, Anti-PI3-kinase p101 subunit, Anti-PI3-kinase regulatory subunit 5, Anti-Phosphoinositide 3-kinase regulatory subunit 5, Anti-PtdIns-3-kinase p101

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PI3KR5(23533)

General description

PI3KR5 is a regulatory subunit of the PI3K gamma complex. This protein, which interacts with G beta gamma proteins, is a heterodimer of a catalytic subunit (PI3KCG/p120) and a regulatory (PI3KR5a/p101) subunit.

Immunogen

PI3KR5 (NP_055123, 397-431)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human PI3KR5.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service