Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300390

Sigma-Aldrich

Anti-ADCK5 (C-term) antibody produced in rabbit

saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution

Synonym(s):

Anti-ADCK5, Anti-AarF domain containing kinase 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

saturated ammonium sulfate (SAS) precipitated

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADCK5(203054)

General description

ADCK5 is member of the AarF domain containing kinase family.
The gene ADCK5 (aarF domain containing kinase 5) is mapped to human chromosome 8q24.3. It is a putative atypical kinase. ADCK5 might be involved in the coenzyme Q biosynthetic pathway.

Immunogen

ADCK5 (NP_777582, )
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human ADCK5.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coenzyme Q biosynthesis in health and disease.
Acosta MJ, et al.
Biochimica et Biophysica Acta, 1857, 1079-1085 (2016)
Genetic bases and clinical manifestations of coenzyme Q10 (CoQ 10) deficiency.
Desbats MA, et al.
Journal of inherited metabolic disease, 38, 145-156 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service