Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300383

Sigma-Aldrich

Anti-FN3K (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Fructosamine-3-kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN3K(64122)

General description

FN3K catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines followed by Amadori rearrangement. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins. Human and mouse FN3K share 86% amino acid identity. GENE Purified FN3K catalyzes ATP-dependent phosphorylation of a synthetic fructosamine, 1-deoxy-1-morpholinofructose (DMF). Recombinant mouse and human FN3K, expressed in E. coli, catalyze phosphorylation of DMF, fructoselysine, fructoseglycine, and fructose in order of decreasing affinity. They also phosphorylate glycated lysozyme, but not unmodified lysozyme. In addition to ATP, FN3K can utilize GTP, CTP, and UTP as phosphate donors.

Immunogen

FN3K (NP_071441, 284-319)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human FN3K.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service