Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300377

Sigma-Aldrich

Anti-FN3X (center) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-FN3KRP, Anti-Fructosamine-3-kinase-related protein, Anti-Ketosamine-3-kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FN3X(79672)

General description

Mammalian fructosamine-3-kinase (FN3K) catalyzes phosphorylation of fructosamines formed by glycation, the nonenzymatic reaction of glucose with primary amines. FN3K for example catalyzes transformation of fructoselysine (FL) residues on glycated proteins, to FL-3-phosphate (FL3P). This phosphorylation destablilizes FL and propels spontaneous decomposition, thereby reversing at an early stage the nonenzymatic glycation process. Phosphorylation of fructosamines may initiate metabolism of the modified amine and result in deglycation of glycated proteins. In mammals, FN3K has a closely related homologue, FN3KRP (FN3K-related protein), which in the human genome is located just 8 kb upstream of FN3K on chromosome 17q25.3 FN3KRP, which is 68% sequence identity with FN3K, is a deglycating enzyme which phosphorylates the third carbon of the sugar moiety of ketosamines, and may play a role in freeing proteins from ribulosamines or psicosamines. A deglycation system may be an important factor in protecting cells from the deleterious effects of nonenzymatic glycation, which appears to be an important factor in the pathogenesis of diabetic complications.

Immunogen

FN3X (Q9HA64, 121-156)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the center region of human FN3X.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service