Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB1300361

Sigma-Aldrich

Anti-MTMR6 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:50-1:100
indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTMR6(9107)

General description

The myotubularin (MTM) family constitutes one of the largest and most highly conserved protein-tyrosine phosphatase (PTP) subfamilies. Myotubularins, contain the consensus active site of tyrosine phosphatases but otherwise shows no homology to other phosphatases. PTP′s usually act on substrates containing only phosphotyrosine sites, but myotubularins were shown to act on both phosphotyrosine and phosphoserine (dual specific). The enzymatic activity of myotubularins had not been demonstrated previously because it lacks catalytically active residues in tyrosine phosphatase/dual-specific phosphatase active site. The active site is however sufficiently preserved to bind phosphorylated substrates, and may protect from phosphatases. It was reported that interaction of myotubularin family members makes one of them catalytically active. The 4.8-kb MTMR6 mRNA is expressed ubiquitously. By analysis of radiation and somatic cell hybrids, the MTMR6 gene has been mapped to 13q12. MTMR6 corresponds to an EST located within a cloned region that encompasses a translocation breakpoint t(8;13) observed in 2 patients with an atypical myoproliferative disorder.

Immunogen

MTMR6 (Q9Y217, 582-617)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human MTMR6.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service