Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300360

Sigma-Aldrich

Anti-MTMR5 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Myotubularin-related protein 5, Anti-SET binding factor 1, Anti-Sbf1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTMR5(6305)

General description

The myotubularin (MTM) family constitutes one of the largest and most highly conserved protein-tyrosine phosphatase (PTP) subfamilies. Myotubularins, contain the consensus active site of tyrosine phosphatases but otherwise shows no homology to other phosphatases. PTP′s usually act on substrates containing only phosphotyrosine sites, but myotubularins were shown to act on both phosphotyrosine and phosphoserine (dual specific). The enzymatic activity of myotubularins had not been demonstrated previously because it lacks catalytically active residues in tyrosine phosphatase/dual-specific phosphatase active site. The active site is however sufficiently preserved to bind phosphorylated substrates, and may protect from phosphatases. It was reported that interaction of myotubularin family members makes one of them catalytically active. MTMR5 can interact with MTMR2 and regulates the enzymatic activity and the subcellular localization of MTMR2. MTMR5 inhibits myoblast differentiation in vitro and induces oncogenic transformation in fibroblasts.Other suggested roles for MTMR5 are in spermatogenesis and germ cell differentiation. Binding partners for MTMR5 include the SET domain of MLL/HRX and SUV39H1.

Immunogen

MTMR5 (1836-1871)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human MTMR5.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service