Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300357

Sigma-Aldrich

Anti-MTMR1 (N-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$455.00

$455.00


Check Cart for Availability


Select a Size

Change View
100 μG
$455.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$455.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTMR1(53332)

General description

MTM1 gene mutations cause X-linked myotubular myopathy. The corresponding protein, myotubularin, contains the consensus active site of tyrosine phosphatases (PTP) and is a tyrosine/serine phosphatase. The 3.7-kb MTMR1 mRNA is expressed ubiquitously. An additional 3.1-kb transcript was detected only in placenta. Analysis of the genomic region containing MTM1 and MTMR1 reveals that the 2 genes share a similar structure, suggesting that they are related and arose from an intrachromosomal gene duplication. The 2 main MTMR1 protein muscular isoforms, like myotubularin, dephosphorylate PI(3)P in vitro. There is a striking reduction in the level of the muscle-specific isoform and the appearance of an abnormal MTMR1 transcript in cultured differentiated muscle cells and in skeletal muscle from congenital myotonic dystrophy patients. MTMR1 may play a role in muscle formation, and may represent another target for abnormal mRNA splicing in myotonic dystrophy.

Immunogen

MTMR1 (Q9Z2C4, 6-42)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MTMR1.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service