MLLT2 is involved in acute leukemias through a chromosomal translocation t(4;11)(q21;q23) that involves mllt2 and mll/hrx. AF-4 (MLLT2), AF-9, and ENL proteins contain nuclear targeting sequences as well as serine-rich and proline-rich regions. Stretches abundant in basic amino acids are also present in the three proteins. These results suggest that the different proteins fused to ALL-1 polypeptide(s) provide similar functional domains. AF4 is a serine- and proline-rich putative transcription factor with a glutamine-rich carboxyl terminus. The composition of the complete MLL-AF4 fusion product argues that it may act through either a gain-of-function or a dominant negative mechanism in leukemogenesis.
Immunogen
MLLT2 (NP_005926, 6-38) This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the N-terminal region of human MLLT2.
Physical form
Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.