Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300330

Sigma-Aldrich

Anti-HDCMC04P antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KMT2E, Anti-Myeloid/lymphoid or mixed-lineage leukemia 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLL5(55904)

General description

This protein has been identified only at the translational level. It is 96% homologous to human MLL5. This polyclonal antibody was raised against a peptide corresponding to a peptide sequence selected from the C-terminal region of HDCMC04P.

Immunogen

HDCMC04P (AAF75564, 429-463)
This polyclonal antibody was raised against a peptide corresponding to a peptide sequence selected from the C-terminal region of HDCMC04P.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service