Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB1300327

Sigma-Aldrich

Anti-MLL2 (C-term) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-KMT2B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: 1:1000

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLL2(8085)

General description

The SET domain is a conserved C-terminal domain that characterizes proteins of the MLL family, including MLL2. The MLL SET domain is a histone H3 Lys4 (K4)-specific methyltransferase whose activity is stimulated with acetylated H3 peptides. The gene for MLL2 encodes a 5,262-amino acid protein containing a SET domain, 5 PHD fingers, potential zinc fingers, and a long run of glutamines interrupted by hydrophobic residues (mostly leucine). They also detected an alternatively spliced form encoding 4,957 amino acids and lacking an N-terminal zinc finger and PHD finger. By analysis of rodent/human hybrid cells and analysis of the Genebridge radiation hybrid panel, they mapped the gene to the 12p13.1-qter region. The 12q12-q13 region is involved in duplications and translocations associated with cancer. By database searching, Karlin et al. (2002) identified 192 human protein sequences that have multiple amino acid runs, many of which are associated with disease, including cancer. Karlin et al. (2002) found that a key aspect of 82 of these protein sequences is their role in transcription, translation, and developmental regulation. MLL2 is a striking example of proteins with multiple amino acid runs, with 22 glutamine runsGenes encoding a significant number of long amino acid runs are potentially associated with diseases, such as cancer.

Immunogen

MLL2 (NP_003473, 4710-4744)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of human MLL2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service